5-(4-Fluorphenyl)isoxazol

Lieferant: Apollo Scientific
Achtung

PC9962-1G PC9962-25G PC9962-5G
APOSPC9962-1GEA 65.5 EUR
APOSPC9962-1G APOSPC9962-25G APOSPC9962-5G
5-(4-Fluorphenyl)isoxazol
5-(4-Fluorophenyl)isoxazole
Formel: C₉H₆FNO
Molecular Weight: 163,15 g/mol
Schmelzpunkt: 53…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183544
CAS-Nummer: 138716-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR