β-Nicotinamide adenine dinucleotide ≥97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Coenzyme I, β-NAD, Diphosphopyridine dinucleotide

BIB3011-1G BIB3011-5G
APOSBIB3011-1GEA 21.9 EUR
APOSBIB3011-1G APOSBIB3011-5G
β-Nicotinamide adenine dinucleotide ≥97%
β-Nicotinamide adenine dinucleotide
Formel: C₂₁H₂₇N₇O₁₄P₂
Molecular Weight: 663,43 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036253
CAS-Nummer: 53-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR