Tobramycin sulphate

Lieferant: Apollo Scientific
Gefahr

BIT0153-1G BIT0153-500MG BIT0153-5G
APOSBIT0153-1GEA 72.5 EUR
APOSBIT0153-1G APOSBIT0153-500MG APOSBIT0153-5G
Tobramycin sulphate
Tobramycin sulphate

Tobramycin is an aminoglycoside antibiotic and hasa bactericidal action against many gram-negative bacteria.

Formel: (C₁₈H₃₇N₅O₉)₂·5H₂SO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00133864
CAS-Nummer: 79645-27-5
EINECS: 256-499-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR