Photometric tube tests, NANOCOLOR® Nitrate
Lieferant: MACHEREY-NAGEL
985064
985066
985065
MANA985064EA
99.1
EUR
MANA985064
MANA985066
MANA985065
Photometric tube tests, NANOCOLOR® Nitrate
Umwelt-Prüfkits
A pre-dosed NANOCOLOR® tube test for photometric determination of nitrate in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.