Hibifolin

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,3S,4S,5R,6S)-6-((2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4-oxo-4H-chromen-8-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid

BIDF1029-10MG BIDF1029-25MG
APOSBIDF1029-10MGEA 197 EUR
APOSBIDF1029-10MG APOSBIDF1029-25MG
Hibifolin
Hibifolin
Formel: C₂₁H₁₈O₁₄
Molecular Weight: 494,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28987619
CAS-Nummer: 55366-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR