Imipenem and cilastatin sodium
Lieferant: Apollo Scientific
|
Achtung
|
BIIC0001-1G
BIIC0001-500MG
BIIC0001-100MG
APOSBIIC0001-1GEA
422
EUR
APOSBIIC0001-1G
APOSBIIC0001-500MG
APOSBIIC0001-100MG
Imipenem and cilastatin sodium
Imipenem and cilastatin sodium
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD01744417 CAS-Nummer: 85960-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...