Amastatin

Lieferant: Apollo Scientific
Achtung

BIMI3764-1MG BIMI3764-5MG
APOSBIMI3764-1MGEA 40.6 EUR
APOSBIMI3764-1MG APOSBIMI3764-5MG
Amastatin
Amastatin
Formel: C₂₁H₃₈N₄O₈
Molecular Weight: 474.548 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038400
CAS-Nummer: 67655-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR