Arabinofuranosyl-2-fluoroadenine ≥99%
Lieferant: Apollo Scientific
Synonyme:
2-Fluoroadenine Arabinosid, (2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, Fludarabine
BIMN2039-1G
BIMN2039-250MG
APOSBIMN2039-1GEA
766
EUR
APOSBIMN2039-1G
APOSBIMN2039-250MG
Arabinofuranosyl-2-fluoroadenine ≥99%
Arabinofuranosyl-2-fluoroadenine
Formel:
C₁₀H₁₂FN₅O₄ Molecular Weight: 285.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00132942 CAS-Nummer: 21679-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...