Arabinofuranosyl-guanine hydrate ≥99%
Lieferant: Apollo Scientific
Synonyme:
Guanine 9-β-D-Arabinofuranoside, Araguanosine, Ara-G
BIMN2040-1G
BIMN2040-250MG
APOSBIMN2040-1GEA
256
EUR
APOSBIMN2040-1G
APOSBIMN2040-250MG
Arabinofuranosyl-guanine hydrate ≥99%
Arabinofuranosyl-guanine hydrate
Formel:
C₁₀H₁₃N₅O₅ Molecular Weight: 283,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065486 CAS-Nummer: 38819-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...