ACTH

Lieferant: Apollo Scientific
Gefahr

BITH3003-10MG BITH3003-2MG BITH3003-50MG
APOSBITH3003-10MGEA 197 EUR
APOSBITH3003-10MG APOSBITH3003-2MG APOSBITH3003-50MG
ACTH
ACTH
Lagertemperatur: Gefrierschrank
MDL: MFCD00145721
CAS-Nummer: 9002-60-2
EINECS: 232-659-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR