DDAVP

Lieferant: Apollo Scientific
Gefahr

BITH3012-100MG BITH3012-25MG BITH3012-5MG
APOSBITH3012-100MGEA 0 EUR
APOSBITH3012-100MG APOSBITH3012-25MG APOSBITH3012-5MG
DDAVP
DDAVP
Lagertemperatur: Gefrierschrank
MDL: MFCD00080163
EINECS: 240-726-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR