Virginiamycin complex

Lieferant: Apollo Scientific

BIV1005-100MG BIV1005-250MG BIV1005-1G
APOSBIV1005-100MGEA 125 EUR
APOSBIV1005-100MG APOSBIV1005-250MG APOSBIV1005-1G
Virginiamycin complex
Virginiamycin complex
Schmelzpunkt: 165-175 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 121006-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR