5-(1,2,3,6-Tetrahydropyridin-4-yl)pyrimidine dihydrochloride

Lieferant: Apollo Scientific

OR321506-1G OR321506-250MG
APOSOR321506-1GEA 957 EUR
APOSOR321506-1G APOSOR321506-250MG
5-(1,2,3,6-Tetrahydropyridin-4-yl)pyrimidine dihydrochloride
5-(1,2,3,6-Tetrahydropyridin-4-yl)pyrimidine dihydrochloride
Formel: C₉H₁₁N₃.₂ClH
Molecular Weight: 234,126 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1864014-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR