1,2,3,6-Tetrahydro-2,3'-bipyridine dihydrochloride

Lieferant: Apollo Scientific

OR321515-1G OR321515-250MG
APOSOR321515-1GEA 877 EUR
APOSOR321515-1G APOSOR321515-250MG
1,2,3,6-Tetrahydro-2,3'-bipyridine dihydrochloride
1,2,3,6-Tetrahydro-2,3'-bipyridine dihydrochloride
Formel: C₁₀H₁₂N₂.₂ClH
Molecular Weight: 233.138 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1347744-97-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR