1-(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methanamine dihydrochloride 95%
Lieferant: Apollo Scientific
Achtung
OR475193-1G OR475193-250MG
APOSOR475193-1GEA 719 EUR
APOSOR475193-1G APOSOR475193-250MG
1-(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methanamine dihydrochloride 95%
1-(4-Bromo-1-cyclopropyl-1H-pyrazol-3-yl)methanamine dihydrochloride
Formel: C₇H₁₀BrN₃.₂ClH
Molecular Weight: 289 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32669859
CAS-Nummer: 2514952-96-4