[(1-Cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride 95%
Lieferant: Apollo Scientific
Achtung
OR475195-1G OR475195-250MG
APOSOR475195-1GEA 719 EUR
APOSOR475195-1G APOSOR475195-250MG
[(1-Cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride 95%
[(1-Cyclopropyl-1H-pyrazol-3-yl)methyl](methyl)amine dihydrochloride
Formel: C₈H₁₃N₃.₂ClH
Molecular Weight: 224.131 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 2514953-06-9