3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific

PC303504-1G PC303504-5G
APOSPC303504-1GEA 0 EUR
APOSPC303504-1G APOSPC303504-5G
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzoic acid
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₂BrCl₂F₃O₂
Molecular Weight: 337,905 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640097
CAS-Nummer: 2514730-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR