4,5,6-Pyrimidintriaminsulfat ≥98%, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Pyrimidine-4,5,6-triamine sulphate
M-Clarity™ Qualitätsstufe = MQ100
307181-1G
307181-5G
SIAL307181-1GEA
99.6
EUR
SIAL307181-1G
SIAL307181-5G
4,5,6-Pyrimidintriaminsulfat ≥98%, Sigma-Aldrich®
4,5,6-Pyrimidintriaminsulfat
4,5,6-Triaminopyrimidine sulfate was converted to 4,5,6-triaminopyrimidine sulfite by formylation with C13 labeled formic acid that was used in the preparation of adenine labeled with C13.
Formel:
C₄H₇N₅·H₂SO₄ Molecular Weight: 223,21 g/mol |
MDL:
MFCD00012789 CAS-Nummer: 6640-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...