Cis-reporting systems, PathDetect
Lieferant: STRATAGENE
219059 219071 219074 219080 219082 219083 219084 219085 219087 219089 219090 219091 240039
STRA219059EA 669 EUR
STRA219059 STRA219071 STRA219074 STRA219080 STRA219082 STRA219083 STRA219084 STRA219085 STRA219087 STRA219089 STRA219090 STRA219091 STRA240039
Cis-reporting systems, PathDetect
Mikroarray-Reagenzien
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
Bitte beachten For research use only. Not for use in diagnostic procedures.