Trans-reporting systems, PathDetect
Lieferant: STRATAGENE
219000 219001 219002 219004 219005 219015 219026 219031 219036 219050 219053 219054 219056 219062 219065 219068 219086 219040 219041 219042
STRA219000EA 1430 EUR
STRA219000 STRA219001 STRA219002 STRA219004 STRA219005 STRA219015 STRA219026 STRA219031 STRA219036 STRA219050 STRA219053 STRA219054 STRA219056 STRA219062 STRA219065 STRA219068 STRA219086 STRA219040 STRA219041 STRA219042
Trans-reporting systems, PathDetect
Mikroarray-Reagenzien
PathDetect trans-reporting systems allow you to determine if the protein or compound of interest directly or indirectly activates one of six transcription factors: c-Jun, Elk1, CREB, c-Fos, ATF2 or CHOP.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
Easily investigate whethe the protein or compound influences the activation of any of the six transcription factors offered and etermine the direct or indirect involvement of new gene products, growth factors, and drug candidates in specific pathways.
Lieferumfang: Contains of a reporter plasmid, a trans-activator plasmid and both positive and negative control plasmids.
Bitte beachten For research use only. Not for use in diagnostic procedures.