Terrestrial (Cypripedium) orchid medium
Lieferant: Apollo Scientific
PMT839A-10X1L PMT839B-10L PMT839C-50L
APOSPMT839A-10X1LEA 46.8 EUR
APOSPMT839A-10X1L APOSPMT839B-10L APOSPMT839C-50L
Terrestrial (Cypripedium) orchid medium
Zellkulturmedien
Contains 400 mg/L calcium nitrate and 400 mg/L Casein; WITHOUT ammonium nitrate.