Potassium hexacyanoruthenate(II) hydrate, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Potassium hexacyanoruthenate hydrate

M-Clarity™ Qualitätsstufe = MQ100

378232-2.5G 378232-500MG
SIAL378232-2.5GEA 415 EUR
SIAL378232-2.5G SIAL378232-500MG
Potassium hexacyanoruthenate(II) hydrate, Sigma-Aldrich®
Potassium hexacyanoruthenate(II) hydrate

Used for the synthesis of ruthenium purple (RP) films on transmissive tin-doped indium oxide (ITO)/glass substrates.

Formel: C₆K₄N₆Ru·xH₂O
Molecular Weight: 413,57 g/mol (anhy)
MDL: MFCD00150398
CAS-Nummer: 339268-21-2
UN: 1588
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR