qPCR Consumables
Lieferant: STRATAGENE
401266 410022 410024
STRA401266EA 124 EUR
STRA401266 STRA410022 STRA410024
qPCR Consumables
PCR-Cycler
These QPCR Plastics and accessories have been validated for optimal performance with Mx3005P, Mx3000P and the AriaMx real time PCR Systems.
Plates and tubes fit most PCR instruments. Racks hold standard PCR and microcentrifuge tubes.
The StrataCooler LP benchtop cooler provides the most protection for enzymes and the cooler will maintain a temperature measuring as low as –15 °C even after 2 hours.
StrataCooler Cryo preservation modules enable highly controlled freezing (0,4 to 0,6 °C/minute) of mammalian and insect cells to significantly increase survival rates (up to 90%).
96-Well Semi-skirted Polypropylene PCR plate validated for optimal performance and compatible with the optical Strip Caps.
Bitte beachten For research use only. Not for use in diagnostic procedures.