Multi-test verification materials, MTVM for the petroleum industry
Lieferant: STANHOPE SETA
99851-0 99853-0 99850-0 99852-0 99854-0 99908-0
751-0044EA 0 EUR
751-0044 751-0046 751-0043 751-0045 751-0047 751-0079
Multi-test verification materials, MTVM for the petroleum industry
Standards Mineralölindustrie-Standards
Multi Test Verification Materials (MTVM) allow routine monitoring of instrument performance and are also particularly helpful in training personnel on new equipment and test methods.
  • Verification to ASTM/CEN/ISO/IP and equivalent test procedures
  • Cross-checks instrument performance using real product