GN25

Lieferant: Cayman Chemical

Synonyme: 3-​[(1,​4-​dihydro-​5,​8-​dimethoxy-​1,​4-​dioxo-​2-​naphthalenyl)thio]-​propanoic acid, p53-Snail binding Inhibitor

10948-5 10948-10 10948-25 10948-50
CAYM10948-5EA 46 EUR
CAYM10948-5 CAYM10948-10 CAYM10948-25 CAYM10948-50
GN25
GN25

A 2-thio-dimethoxy naphthoquinone analog that blocks Snail binding to p53 and induces p53 expression in cancer cells in a K-Ras dependent manner

Formel: C₁₅H₁₄O₆S
Molecular Weight: 322.3 g/mol
CAS-Nummer: 1227401-27-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR