SynaptoGreen™ C5 (Equivalent to FM®1-84)
Lieferant: Biotium
70046 70047
70046.EA 442 EUR
70046. 70047.
SynaptoGreen™ C5 (Equivalent to FM®1-84)
SynaptoGreen™ C5 (Equivalent to FM®1-84)
SynaptoGreen™ C5 is similar to SynaptoGreen™ C4 (70020), except that the lipophilic tail is one carbon longer.
  • Ex/Em: 510/625 nm (in MeOH)
  • Red solid soluble in water
SynaptoGreen™ C5 (equivalent to FM®1-84) is similar to SynaptoGreen™ C4 (70020), except that the lipophilic tail is one carbon longer. Thus SynaptoGreen™ C5 is less water soluble than SynaptoGreen™ C4 and is predicted to have a faster staining rate but slower de-staining rate.