HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure), Pulver säurefrei, Corning®
Lieferant: Corning
Synonyme: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
61-034-RR 61-034-RO 61-034-RM
MDTC61-034-RREA 607.86 EUR
MDTC61-034-RR MDTC61-034-RO MDTC61-034-RM
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure), Pulver säurefrei, Corning®
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
HEPES (N-2-hydroxyethylpiperazine-N’-2-ethanesulphonic acid) provides an alternative to the use of bicarbonate for buffering because it does not require elevated levels of CO2
Unlike the low pKa of bicarbonate, HEPES has a pKa of 7.66 making it a much stronger buffer in the pH 7.2-7.6 range than bicarbonate buffer. Potential disadvantages of HEPES include toxicity over 100 mM and lack of nutritive value to the cell.
Formel: C₈H₁₈N₂O₄S
Molecular Weight: 238.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006158
CAS-Nummer: 7365-45-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM