Human IL-1 alpha / IL1A / IL1F1 protein
Lieferant: Sino Biological
10128-HNCH-100 10128-HNCH-20
SINO10128-HNCH-100EA 0 EUR
SINO10128-HNCH-100 SINO10128-HNCH-20
Human IL-1 alpha / IL1A / IL1F1 protein
Proteine und Peptide
The processed form of human IL1α (NP_000566.3) (Ser 113-Ala 271) was expressed and purified.