Clobetasolpropionat ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cloβsol propionate, Clobetasol-17-propionate, 21-Chloro-9ɑ-fluoro-11β,17ɑ-dihydroxy-16β-methyl-1,4-pregnadiene-3,20-dione 17-Propionate

466820010 466820050
ACRO466820010EA 46.3 EUR
ACRO466820010 ACRO466820050
Clobetasolpropionat ≥97%
Clobetasol propionate
Formel: C₂₅H₃₂ClFO₅
Molecular Weight: 466,97 g/mol
MDL: MFCD00058499
CAS-Nummer: 25122-46-7
EINECS: 246-634-3

Bestellen

Spezifikation Testergebnisse

Appearance Crystalline powder, White to almost white
Infrared spectrum Conforms
Melting point 196.0°C to 200.0°C
HPLC >=97.0 %
Loss on drying =<0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR