3-Hydroxytyrosol

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol

467630010 467630050
ACRO467630010EA 172 EUR
ACRO467630010 ACRO467630050
3-Hydroxytyrosol
3-Hydroxytyrosol
Formel: C₈H₁₀O₃
Molecular Weight: 154.17 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01320529
CAS-Nummer: 10597-60-1

Bestellen

Spezifikation Testergebnisse

Appearance Liquid, Clear colorles to yellow
Infrared spectrum Conforms
Refractive index 1.5810 to 1.5840 (20°C, 589 nm)
HPLC >=97.5 %
Water =<5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR