Mouse EpCAM / TROP-1 / TACSTD1 Protein (ECD, His Tag)
Lieferant: Sino Biological
50591-M08H 50591-M08H-100
SINO50591-M08HEA 0 EUR
SINO50591-M08H SINO50591-M08H-100
Mouse EpCAM / TROP-1 / TACSTD1 Protein (ECD, His Tag)
Proteine und Peptide
A DNA sequence encoding the extracellular domain of mouse EpCAM (NP_032558.2) (Met 1-Thr 266) was expressed, with a polyhistidine tag at the C-terminus.