Cynomolgus IL13 Protein
Lieferant: Sino Biological
11057-CNAH-100
11057-CNAH-1
SINO11057-CNAH-100EA
561
EUR
SINO11057-CNAH-100
SINO11057-CNAH-1
Cynomolgus IL13 Protein
Proteine und Peptide
A DNA sequence encoding the cynomolgus (Macaca fascicularis) IL13 (ABG75889.1) (Met 1-Asn 132) precursor was expressed.