Natriumoleat ≥95%
Lieferant: Cayman Chemical
Synonyme: cis-9-Octadecensäuremethylester Natriumsalz, Ölsäure Natriumsalz
24659-100 24659-25 24659-50 24659-10
CAYM24659-100EA 464 EUR
CAYM24659-100 CAYM24659-25 CAYM24659-50 CAYM24659-10
Natriumoleat ≥95%
Natriumoleat
Oleic acid is a monounsaturated fatty acid and a major component of membrane phospholipids that has been found in human plasma, cell membranes, and adipose tissue.
Bitte beachten This product is not for human or veterinary use.
Formel: C₁₈H₃₃NaO₂
Molecular Weight: 304.45 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00004438
CAS-Nummer: 143-19-1