Prostaglandin A1 ≥98%
Lieferant: Cayman Chemical
Synonyme:
PGA1, 9-oxo-15S-hydroxy-prosta-10,13E-dien-1-oic acid
10010-1
10010-10
10010-5
10010-50
CAYM10010-1EA
48
EUR
CAYM10010-1
CAYM10010-10
CAYM10010-5
CAYM10010-50
Prostaglandin A1 ≥98%
Prostaglandin A1
Prostaglandin A1 (PGA1) was first isolated as a dehydration product of the PGE1 compounds found in human semen.It has been shown to cause renal vasodilation, increased urine sodium excretion, and lowered arterial pressure in hypertensive patients.2 It also has growth-inhibitory activity on tumor cells; the IC50 value for the inhibition of human ovarian cancer is 7.5 µM.
Formel:
C₂₀H₃₂O₄ Molecular Weight: 336,5 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
14152-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...