3-(4-Hydroxyphenyl)-7-chromanol ≥99,0% (durch TLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Hydroxyphenyl)chroman-7-ol, (R,S)-Equol, 4′,7-Dihydroxyisoflavane, 7-Hydroxy-3-(4-hydroxyphenyl)chroman, (±)-Equol, 4',7-Isoflavandiol, 3-(4-Hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol

M-Clarity™ Qualitätsstufe = MQ100

45405-1MG 45405-5MG
SIAL45405-1MGEA 482 EUR
SIAL45405-1MG SIAL45405-5MG
3-(4-Hydroxyphenyl)-7-chromanol ≥99,0% (durch TLC), Sigma-Aldrich®
(+/-)-Equol

Metabolite of daidzein. Weak estrogenic agonist, and competitive inhibitor of 17β-estradiol at the estradiol receptor.

Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
MDL: MFCD00016662
CAS-Nummer: 94105-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR