Fipronil

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-3-cyano-4-(trifluoromethyl)sulphinylpyrazole, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)sulphinylpyrazole-3-carbonitrile

465570050 465570250 465571000
ACRO465570050EA 57.5 EUR
ACRO465570050 ACRO465570250 ACRO465571000
Fipronil
Fipronil
Formel: C₁₂H₄Cl₂F₆N₄OS
Molecular Weight: 437,14 g/mol
Schmelzpunkt: 200°C…201°C
Lagertemperatur: Kühlschrank
MDL: MFCD00867812
CAS-Nummer: 120068-37-3
UN: 2811
ADR: 6.1,III
Merck Index: 13.4115

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige powder or crystals
Infrared spectrum Conforms
Melting point 194°C to 205°C
HPLC ≥ 97.5%
Loss on drying ≤ 2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR