Tenofovir

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-(((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphonic acid, (R)-[[2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid

465610010 465610050
ACRO465610010EA 56 EUR
ACRO465610010 ACRO465610050
Tenofovir
Tenofovir
Formel: C₉H₁₄N₅O₄P
Molecular Weight: 287.22 g/mol
Schmelzpunkt: 283°C
Lagertemperatur: Gefrierschrank
MDL: MFCD00943794
CAS-Nummer: 147127-20-6

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white powder
Infrared spectrum Conforms
Water ≤ 3%
HPLC ≥ 97.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR