4-Pyridoxic acid 98, Solid
Lieferant: Cayman Chemical
|
Achtung
|
Synonyme:
3-hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxylic acid, 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinic acid
4-Pyridoxic acid is an inactive metabolite of pyridoxine. It is formed from pyridoxine via pyridoxal 5'-phosphate and pyridoxal intermediates by pyridoxine kinase or pyridoxamine phosphate oxidase, as well as pyridoxine-5'-phosphate oxidase, aldehyde dehydrogenase, or aldehyde oxidase.
Solubility: Aqueous acid: 10 mg/ml
Bitte beachten This product is not for human or veterinary use.
Formel:
C₈H₉NO₄ Molecular Weight: 183.2 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006334 CAS-Nummer: 82-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...