Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ohira-Bestmann Reagent, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, (1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester
466020010
466020050
ACRO466020010EA
99.8
EUR
ACRO466020010
ACRO466020050
Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Dimethyl (1-diazo-2-oxopropyl)phosphonate
Formel:
C₅H₉N₂O₄P |
MDL:
MFCD07368360 CAS-Nummer: 90965-06-3 |
Spezifikation Testergebnisse
Appearance (Color) | Clear light yellow to brown |
Appearance (Form) | Liquid |
Refractive index | 1.4770 to 1.4820 (20°C, 589 nm) |
HPLC | >=94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...