Echtgrün FCF
Lieferant: Spectrum Chemical
Achtung
Synonyme: Dihydrogen(ethyl)[4-[4-[ethyl(3-sulfonatobenzyl)amino](4-hydroxy-2-sulfonatobenzhydryliden]cyclohexa-2,5-dien-1-yliden](3-sulfonatobenzyl)ammonium Dinatriumsalz, Fast Green FCF
FD120-100GM FD120-500GM
SPCMFD120-100GMEA 459 EUR
SPCMFD120-100GM SPCMFD120-500GM
Echtgrün FCF
Echtgrün FCF
FD and C Green No. 3 | Non-graded FD and C Green No. 3 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel: C₃₇H₃₄N₂Na₂O₁₀S₃
Molecular Weight: 808,86 g/mol
Schmelzpunkt: 290 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013053
CAS-Nummer: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970