Eisen(II)sulfat Heptahydrat 99,5, granular USP
Lieferant: Spectrum Chemical
Synonyme: Eisenvitriol
FE110-500GM FE110-2.5KG
SPCMFE110-500GMEA 151 EUR
SPCMFE110-500GM SPCMFE110-2.5KG
Eisen(II)sulfat Heptahydrat 99,5, granular USP
Eisen(II)sulfat Heptahydrat
Ferrous Sulfate, Heptahydrate, Granular, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel: FeSO₄·7H₂O
Molecular Weight: 278.02 g/mol
Schmelzpunkt: 64
Lagertemperatur: Raumtemperatur
MDL: MFCD00149719
CAS-Nummer: 7782-63-0
Merck Index: 13,04091
Bestellen
Spezifikation Testergebnisse
Assay (As Heptahydrate) 99.5 - 104.5 %
Arsenic max.3 ppm
Lead max. 10 ppm
Mercury max. 3 Ug/G
Identification To Pass Test