Benzoylperoxid 75%, wasserhaltig ≥75% USP
Lieferant: Spectrum Chemical
Synonyme: Dibenzoyl peroxide
BE156-10KGBL BE156-1KGBL
SPCMBE156-10KGBLEA 3480 EUR
SPCMBE156-10KGBL SPCMBE156-1KGBL
Benzoylperoxid 75%, wasserhaltig ≥75% USP
Benzoylperoxid
Hydrous Benzoyl Peroxide, 75 Percent, USP is used to treat mild to moderate acne. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Flame;Flame over circle;Exclamation mark
UN3104.
Formel: C₁₄H₁₀O₄
Molecular Weight: 242.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003071
CAS-Nummer: 94-36-0
UN: 3104
ADR: 5.2 II
Bestellen
Spezifikation Testergebnisse
Assay (Benzoyl Peroxide) 70.0 - 80.0 %
Organic Impurities To Pass Test
Identification To Pass Test