Mangan(IV)oxid

Lieferant: Merck

Synonyme: Mangandioxid, Pyrolusit, Braunstein

243442-100G 243442-500G
SIAL243442-100GEA 71.4 EUR
SIAL243442-100G SIAL243442-500G
Mangan(IV)oxid
Mangan(IV)oxid
Formel: MnO₂
Molecular Weight: 86,94 g/mol
MDL: MFCD00003463
CAS-Nummer: 1313-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR