2-Chlorbiphenyl (PCB Nr. 1) ≥97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chlor-2-phenylbenzol, PCB Nr. 1, 2-PCB, 2-Monochlorbiphenyl

OR7032-1G OR7032-5G
APOSOR7032-1GEA 22 EUR
APOSOR7032-1G APOSOR7032-5G
2-Chlorbiphenyl (PCB Nr. 1) ≥97%
2-Chlorbiphenyl (PCB Nr. 1)
Formel: C₁₂H₉Cl
Molecular Weight: 188.66 g/mol
Siedepunkt: 274 °C
Schmelzpunkt: 32…34 °C
Dichte: 1.1499
CAS-Nummer: 2051-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR