Locust bean gum

Lieferant: Thermo Scientific

Synonyme: Locust (carob) bean gum

466670010 466672500
ACRO466670010EA 155 EUR
ACRO466670010 ACRO466672500
Locust bean gum
Locust bean gum
Lagertemperatur: Raumtemperatur
MDL: MFCD00131257
CAS-Nummer: 9000-40-2

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to light brown powder
Infrared spectrum Conforms
Loss on drying =<12%
Residue after ignition =<1.2%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR