VWR®, Combined Six Cation Standard-II, 6 components in H₂O
Lieferant: VWR Chemicals
91291.154EA 133 EUR
91291.154 91291.180
VWR®, Combined Six Cation Standard-II, 6 components in H₂O
Standards Kationischer Standards Mehrelement-Standards
6 components: Lithium (Li+) 50 mg/l, Sodium (Na+) 200 mg/l, Ammonium (NH4+) 250 mg/l, Potassium (K+) 500 mg/l, Calcium (Ca2+) 500 mg/l, Magnesium (Mg2+) 250 mg/l in H₂O
Standards: ISO 17025/17034
Bestellen
TECHNISCHE DATEN
Environmentally Preferable
Lagertemperatur Ambient