6-Bromo-4-cyanoquinoline ≥95%

Lieferant: Apollo Scientific

OR70308-250MG OR70308-1G
APOSOR70308-250MGEA 241 EUR
APOSOR70308-250MG APOSOR70308-1G
6-Bromo-4-cyanoquinoline ≥95%
6-Bromo-4-cyanoquinoline
Formel: C₁₀H₅BrN₂
Molecular Weight: 233.0641 g/mol
MDL: MFCD28669044
CAS-Nummer: 1542874-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR