5-Bromo-4-cyanoquinoline ≥95%

Lieferant: Apollo Scientific

OR70309-250MG OR70309-1G
APOSOR70309-250MGEA 219 EUR
APOSOR70309-250MG APOSOR70309-1G
5-Bromo-4-cyanoquinoline ≥95%
5-Bromo-4-cyanoquinoline
Formel: C₁₀H₅BrN₂
Molecular Weight: 233,0641 g/mol
MDL: MFCD32862640
CAS-Nummer: 2640734-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR