7-Bromo-4-cyanoquinoline-n-oxide ≥95%
Lieferant: Apollo Scientific
OR70312-250MG
OR70312-1G
APOSOR70312-250MGEA
227
EUR
APOSOR70312-250MG
APOSOR70312-1G
7-Bromo-4-cyanoquinoline-n-oxide ≥95%
7-Bromo-4-cyanoquinoline-n-oxide
Formel:
C₁₀H₅BrN₂O Molecular Weight: 249,0635 g/mol |
MDL:
MFCD32862644 CAS-Nummer: 2640734-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...