Ethyl 4-chloro-1-methyl-6-nitro-2-oxoquinoline-3-carboxylate ≥95%
Lieferant: Apollo Scientific
OR71094-250MG
OR71094-1G
APOSOR71094-250MGEA
241
EUR
APOSOR71094-250MG
APOSOR71094-1G
Ethyl 4-chloro-1-methyl-6-nitro-2-oxoquinoline-3-carboxylate ≥95%
Ethyl 4-chloro-1-methyl-6-nitro-2-oxoquinoline-3-carboxylate
Formel:
C₁₃H₁₁ClN₂O₅ Molecular Weight: 310,68984 g/mol |
MDL:
MFCD33395017 CAS-Nummer: 2253790-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...